Learn More
Perkin Elmer US LLC Continuous Wave Module with Upper Bracket - 150 W
Supplier:  Perkin Elmer US LLC N4202035
Encompass_Preferred
Our Fluorescence Spectrometers use Pulsed Xenon Discharge lamps. These reliable, stable lamps provide remarkable sensitivity and can achieve a peak intensity exceeding 1 KW. For optimal instrument perfomance, always use original PerkinElmer replacement lamps.
Catalog No. 50-211-2316