Learn More
Contec™ PROSAT™ Presaturated Wipes in SatBox™ Containers
BIOPHARMA
Reduce solvent usage with presatured wipes. Contec™ PROSAT™ Presaturated Wipes in SatBox™ Containers are ideal for wiping process tools and other equipment, as well as many other cleaning applications in critical environments. The reusable SatBox container is made from high-density polypropylene and reduces solvent evaporation.
Supplier:  Contec™ CSCN0006
 View more versions of this product
Catalog No. 19-084-311
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
This item is not returnable. View return policy
Description
Description
  • Premoistened knitted polyester wipers and nonwoven polypropylene wipes are saturated with various solvent blends
  • Polynit Heatseal wipers are standard weight, textured 100% polyester interlock knit with nonabrasive, heatsealed edges
  • Polyester wipes are abrasion and chemical resistant
  • Economical nonwoven wipes are made of 100% meltblown polypropylene
  • Ideal for wiping process tools and other equipment, as well as many other cleaning applications in critical environments
  • Reusable SatBox container is made from high-density polypropylene and is compatible with various blends of isopropyl alcohol, ethanol, and acetone
  • SatBox reduces solvent evaporation.
Specifications
Specifications
SatBox Container
24.9 cm
19.8 cm
9.8 x 7.8 in. (24.9 x 19.8 cm)
76266-226
9.8 in.
7.8 in.
Plastic
Controlled Environment Cleaning
Product Suggestions
Product Suggestions