Learn More
Contec™ Bucket Wringers and Frames
Use with Contec™ bucket systems and mops. Contec™ Bucket Wringers and Frames are available in three styles to match the selected Contec™ mop system.
Supplier:  Contec™ 2710
 View more versions of this product
Catalog No. 19-159-255
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
Description
Description
Use with Contec™ bucket systems and mops. Contec™ Bucket Wringers and Frames are available in three styles to match the selected Contec™ mop system.
Use with Contec™ bucket systems and mops. Contec™ Bucket Wringers and Frames are available in three styles to match the selected Contec™ mop system.
  • Choose from downpress, flat or curved styles
  • Available in polypropylene or stainless steel
  • Ideal for cleanrooms and controlled environments with small autoclaves or limited storage space
  • Select wringer that complements mop
Curved stainless-steel sieve 19-200-014 for EasyCurve™ mop and flat stainless-steel sieve 19-033-715 for flat mops such as VertiKlean™ MAX and QuickTask. Sieves require stainless-steel frame 19-033-716 for 5 gal. (20 L) buckets and 19-033-717 for 6.5 gal. (25 L) buckets.
Specifications
Specifications
Heavy Duty Downpress Mop Wringer
Reusable
Contec™ Edgeless™ VertiKlean MAX™ or QuickTask™ Mops, 5 gal. (20 L) or 6.5 gal. (25 L) Bucket
Autoclavable
Ideal for cleanrooms and controlled environments
Non-sterile
Product Suggestions
Product Suggestions