Learn More
Uline Container Dri II Desiccants - Cargo Bag, TYVEK BAG, 6 EA
Supplier: Uline S14045
Protects against condensation damage in long distance railroad or cargo shipments, Calcium chloride traps up to 280 of its weight in moisture, Large bags with clip-on hook to hang or suspend in containers railcars and barges, 5 to 6 bags recommended per 40-ft container or box and hopper cars, Non-toxic Can be disposed of with regular waste, Will turn to gel when it has adsorbed all the moisture it can, The more moisture it adsorbs the harder the bags get, Also known as Container Dri II Plus, Place with cargo print side up, Do not place in direct contact with product, Cannot be reused, Adsorbs 280 of its weight in water vapor, Will not dry out a wet container, Desiccants only prevent moisture build up in closed containers
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.