Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fischer Technical Company™ Contact Plates
Contact plates ideal for testing surfaces.
$34.83 - $45.32
Specifications
Format | Contact Plate |
---|---|
Manufacturing Origin | US with Globally Sourced Materials |
Product Type | Contact Plate |
Sterility | Sterile |
Test Type | Microbiological Tests |
Alert:
Prop 65: Cancer and Reproductive Harm.
Catalog Number | Mfr. No. | Media Type | Detects | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Media Type | Detects | Price | Quantity & Availability | ||||
S012400
|
Fischer Technical Company™
CM100RP15 |
Tryptic Soy Agar | Bacteria |
Pack of 10 for $35.20
|
|
||||
S012393
|
Fischer Technical Company™
CM392RP15 |
Sabouraud Dextrose Agar with Lecithin and Polysorbate 80 | Fungi |
Pack of 10 for $45.32
|
|
||||
S012395
|
Fischer Technical Company™
CM163RP15 |
Tryptic Soy Agar with Lecithin and Polysorbate 80 | Microorganisms |
Pack of 10 for $35.20
|
|
||||
S012396
|
Fischer Technical Company™
CM149RP15 |
Potato Dextrose Agar | Bacteria and Fungi |
Pack of 10 for $35.20
|
|
||||
S012398
|
Fischer Technical Company™
CM103RP15 |
Levine EMB Agar | Gram Negative Bacteria |
Pack of 10 for $34.83
|
|
||||
Description
- Available in a variety of formulations
- For environmental monitoring of surfaces
- Steam sterilized
- Agar is slightly raised to make it easy to test surfaces
Specifications
Contact Plate | |
Contact Plate | |
Microbiological Tests | |
10 Plates |
US with Globally Sourced Materials | |
Sterile | |
Microbiological Testing | |
65 x 15 mm |
Safety and Handling
ShelfLife : 90 Days
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title