Learn More
Yamato Scientific America, Inc. Yamato 14L Constant Temperature Water Bath, Fits 6 containers small and large, 115V/220V
Supplier:  Yamato Scientific America, Inc. BS660
Encompass_Preferred
Energy-saving 14L constant temperature water bath with overheat prevention device and electric leakage breaker. Fits small and large containers. Up to 6 pcs. can be used simultaneously. Sink level can be adjusted.
Catalog No. 50-189-8152