Learn More
Pluriselect Connector Ring, single packed, sterile, 25/Pk
Supplier: Pluriselect 415000003
The Connector Ring is a sterile device to support active filtration, cell detachment, cell lysis or cell stimulation in combination with a pluriStrainer and a 50ml standard (conical) centrifuge tube. pluriStrainer, Connector Ring and a 50ml tube can be combined to a unit. This unit allows for a controlled flow through the sieve. When the Luer-lock of the Connector Ring is closed, sample or liquid remains on the mesh. An open Luer-lock starts the separation or the filtration process.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.