Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Pyramex™ Conical Banded Ear Plugs
Click to view available options
Banded; NRR 22 dB
Description
- Designed for environments with on-and-off noise hazards
- Flexible band can rest around neck when not in use
- Conical ear plugs that require no roll down
Specifications
ANSI S3.1974 | Yes |
Cord Color | Black |
Cord Material | Polypropylene |
Size | Universal |
For Use With (Application) | Environments with on-and-Off Noise Hazards |
High Visibility | Yes |
Plug Material | Polyurethane |
Plug Color | Orange |
Noise Reduction Rating (NRR) | 22 dB |
Corded | Corded |
Show More |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction