Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Accuform™ Cone Chain Kit

Accuform™ Cone Chain Kit attaches to traffic cones to create a barrier system for sectioning-off hazards or danger zones quickly, and effectively

Supplier:  Accuform™ FBC110

Catalog No. 19-169-595


Add to Cart

Description

Description

  • Holder has four hooks to attach chain to
  • Inside diameter of holder is 2.75 in.
  • Kit includes 2 in. x 50 ft. long plastic chain and six plastic chain connectors
Specifications

Specifications

Chain Connector Kit
Sectioning Off Hazards, or Danger Zones
Traffic Cones
Long Plastic Chain (2 in. x 50 ft.), and Plastic Chain Connectors (6)
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit