Learn More
Transcat Eil Conductivity Std.Calibration Solution, 447, 1 pint
Supplier: Transcat Eil WD0065347
Oakton Model # WD-00653-47, ±1% Accuracy at 25°C, NIST Traceable materials, Oakton conductivity/TDS calibration solutions are cross checked by using different test methods, premixed and ready to use. Solutions come in 1 pint bottles. Dimensions: 7.75"H x 2.25" dia.