Learn More
Cole-Parmer Oakton Conductivity and TDS Standard, 12,880 µS; 500 mL
Supplier: Cole-Parmer 0060610
Ideal for laboratory and field analysis, all Oakton Conductivity and TDS Standards are traceable to NIST standard reference materials.Their solutions help you calibrate conductivity instruments to ensure accuracy with standard values given at 25C. Use when readings are expected to be around 12.88 mS/cm (12,880 S/cm).