Learn More
Horiba Instruments LAQUAtwin EC-11 Compact Conductivity Meter
Supplier: Horiba Instruments 3999960125
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
This item has a minimum order quantity of 25 per supplier requirements. The Horiba LAQUAtwin EC-11 Compact Conductivity Meter is the only pocket meter that directly measures conductivity and total dissolved solids (TDS) in 0.12ml sample. No need for a beaker to calibrate your meter or measure a sample. Just place few drops of the standard or sample onto the Horiba LAQUAtwin EC-11 sensor. This procedure saves you time and prevents wasting your precious sample.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.