Learn More
Chemglass Life Sciences Condenser, Friedrichs, 330mm, 24/40 Inner Joint
Supplier: Chemglass Life Sciences CG121101
Condenser, Friedrichs, 330mm, 24/40 Inner Joint. Similar to CG-1210 but with the 24/40 outer joint replaced with a serrated hose connection for use with a drying tube. This style condenser is used primarily in Soxhlet extractions or in reflux mode. Hose connections have an O.D. of 10mm at the middle serration.