Learn More
Wilmad Labglass CONDENSER LIEBIG 24/40 300MM
Supplier: Wilmad Labglass LG5150104
Condenser, Liebig. Jacket Length: 300mm; Joint Size: [ST] 24/40. w/ a standard taper Outer joint at the top and a standard taper inner drip tip joint of the same size at the bottom.