Learn More
Chemglass Life Sciences Condenser, Liebig, 185mm, 14/20 Top Outer, 14/20 Lower Inner, 110mm Jacket Length
Supplier: Chemglass Life Sciences CG1218A20
Condenser, Liebig, 185mm, 14/20 Top Outer, 14/20 Lower Inner, 110mm Jacket Length. Small scale Liebig condenser with a standard taper outer joint at the top and a lower inner drip tip joint. Hose connections have an O.D. of 10mm at the middle serration.