Learn More
Ace Glass, Inc. Condenser, Graham, 500mm, 24/40
Supplier: Ace Glass, Inc. 597716
500mm Graham condenser with 24/40 standard taper inner and outer joint at top and bottom. Hose connections are size D for 3/8 ID tubing.