Learn More
Chemglass Life Sciences Condenser, Gooseneck, Medium, 24/40 Inner Joint, Distilling
Supplier: Chemglass Life Sciences CG121221
Condenser, Gooseneck, Medium, 24/40 Inner Joint, Distilling. Unique one-piece distilling condenser is actually a 75º adapter, highly efficient coil condenser and vacuum adapter in a compact one-piece unit. Available with a choice of three sizes of condensers with uptake inner joint as listed. Top thermometer joint is a 10/30 outer for use with a 3in immersion CG-3506 thermometer. Receiver joint on all sizes is a 24/40 inner having a sealed-in drip tip.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.