Learn More
Chemglass Life Sciences Condenser, Friedrichs, 330mm, 24/40 Joint
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Chemglass Life Sciences CG121001
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Condenser, Friedrichs, 330mm, 24/40 Joint. Condenser has a specially formed spiral inner tube providing a long vapor path, ensuring good heat transfer and anti-flooding characteristics. Condenser has a standard taper outer joint at a 15º angle to the condenser and a lower inner drip tip joint. Hose connections have an O.D. of 10mm at the middle serration.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.