Learn More
Chemglass Life Sciences Condenser, Dewar Type, 14/20 Joint, 90 x 37mm
Supplier: Chemglass Life Sciences CG1209A20
Condenser, Dewar Type, 14/20 Joint, 90 x 37mm. Condenser has a large opening at the top for use with dry ice/acetone or liquid nitrogen. Condenser has an outer standard taper joint set at a 75º angle to the condenser body and a lower inner drip tip joint, both being the same size.