Learn More
Chemglass Life Sciences Cold Finger Only, Minum-Ware
SDP
Supplier:  Chemglass Life Sciences MW4302
Encompass_Preferred
Cold Finger Only, Minum-Ware. Cold finger style condenser is made to fit into MW-50 Hickman still head while the lower drip tip directs distillate into the collection collar.
Catalog No. 50-153-2680