Learn More
Chemglass Life Sciences Condenser, Allihn, 730mm, 24/40 Joint, 600mm Jacket Length
Supplier: Chemglass Life Sciences CG120611
Condenser, Allihn, 730mm, 24/40 Joint, 600mm Jacket Length. Allihn style reflux condenser with a standard taper outer joint at the top and a lower inner drip tip joint. Hose connections have an O.D. of 10mm at the middle serration.