Learn More
Wilmad Labglass CONDENSER ALLIHN 24/40 400MM
Supplier: Wilmad Labglass LG4680106
Condenser, Allihn. Jacket Length: 400mm; Joint Size: [ST] 24/40. Allihn Condenser w/ a standard taper Outer joint at the top and a standard taper inner drip tip joint of the same size at the bottom.