Learn More
Ace Glass, Inc. Condenser, long path, pilot plant, 600mm, 24/40
Supplier: Ace Glass, Inc. 601217
600mm long path pilot plant condenser with 24/40 standard taper joints. The turbulent flow created by the condenser is desirable for use under reduced pressure. Internal and external cooling surfaces result in high efficiency per unit length.