Learn More
METTLER TOLEDO COND SENSOR 3/4NPT 0.1C TI/SS2
Supplier: METTLER TOLEDO 58031201
This analog conductivity sensor is a 2-electrode sensor. Due to its titanium concentric electrodes,this sensor is suitable for measurement in ultra-pure and high-purity water.