Learn More
METTLER TOLEDO COND SENSOR 1.5TRI 0.1C SS 55M
Supplier:  METTLER TOLEDO 58031412
Encompass
Cond Sensor 1.5Tri 0.1C SS 55mmISMITEM 580314122-Electrode Conductivity Sensor.The UniCond 2-electrode conductivity sensor provides an exceptionally wide measurement range due to its advanced built-in circuit. The sensor can be used to measure everything from ultrapure water to brackish water (up to 50,000 μS / cm).One Sensor for an Extensive Measurement RangeThe digital UniCond sensor reduces equipment inventory by enabling the measurement of an extensive water purity range with a single sensor model.Superior Measurement AccuracyThe advanced measuring techniques of the UniCond conductivity sensor contribute to superior accuracy over an extended range.High Accuracy in Pure WaterUniCond exceeds performance of conventional sensors due to built-in measuring circuits and analog-to-digital signal conversion in the sensor head.
Catalog No. NC2006310