Learn More
Vector Laboratories Concanavalin A (ConA) Biotinylated 5 mg
Supplier: Vector Laboratories B10055
Concanavalin A (ConA) recognizes α-linked mannose residues, which are present as part of a "core oligosaccharide" in many serum and membrane-bound glycoproteins. This biotinylated ConA conjugate features a ratio of biotin to lectin protein that provides optimal avidin/streptavidin and lectin binding activity. Supplied essentially free of unconjugated biotins, it is preserved with sodium azide.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.