Learn More
Bellco Glass, Inc. Bellco compression fittings are designed using our standard 45mm blue polypropylene open top caps and white silicone gaskets. A wide variety of fittings are available to allow ph and do monitoring via insertion of these probes into sidearms or vessels. Be
Supplier: Bellco Glass, Inc. 196490010
Bellco compression fittings are designed using our standard 45mm blue polypropylene open top caps and white silicone gaskets. A wide variety of fittings are available to allow ph and do monitoring via insertion of these probes into sidearms or vessels. Be