Learn More
Dukal™ Composite Island Dressings
These Composite Island Dressings are waterproof and feature a non-adherent contact layer and non-woven backing with adhesive border to keep wounds dry and contaminants out.
$138.00 - $259.00
Catalog Number | Mfr. No. | Dimensions (L x W) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Dimensions (L x W) | Price | Quantity & Availability | |||||
22-415-745
|
Dukal™
4082 |
4 x 4 in. |
Case of 200 for $196.00
|
|
|||||
22-415-747
|
Dukal™
4085 |
4 x 6 in. |
Case of 100 for $152.00
|
|
|||||
22-415-743
|
Dukal™
4080 |
4 x 10 in. |
Case of 100 for $195.00
|
|
|||||
22-415-744
|
Dukal™
4081 |
4 x 14 in. |
Case of 50 for $138.00
|
|
|||||
22-415-746
|
Dukal™
4083 |
6 x 6 in. |
Case of 100 for $186.00
|
|
|||||
22-415-748
|
Dukal™
4086 |
6 x 8 in. |
Case of 100 for $259.00
|
|
|||||
Description
- Non-adherent contact layer that covers the wound site.
- Features a soft absorbent pad that collects exudate and protects the wound from trauma.
- Non-woven backing with adhesive border that is gentle and holds pad in place.
- Waterproof to keep wounds dry and keep external contaminants out.
Specifications
Sterile | |
Yes |