Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GSC Go Science Crazy Compass Board
SCI_ED

Great for a simple lines of force demonstration.

Supplier:  GSC Go Science Crazy 313510

Catalog No. S24599

Item Discontinued This item has been discontinued and is no longer available. Please reach out to your sales representative.


Description

Description

  • Ungraduated and transparent
  • Size: 20mm diameter 
Specifications

Specifications

Ungraduated
Ship Weight: 3 lb.
Transparent
Compass Board
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit