Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Compact Digital Dry Bath/ Block Heater, Compact Dry Bath D, 100-240V, CN, EU,and UK plug
SCI_ED

Catalog Number S29053
Click to view available options
Temperature Range (Metric):
Ambient +2° to 100°C

Maximize bench space with our compact, lightweight dry bath with multiple combinations of block sizes for versatility.

  • Built-in temperature-sensing probe assists with outstanding temperature accuracy and control
  • Designed for precise temperature control
  • Fast heating rate
  • Multiple combination of different size of blocks allows for maximum tube size versatility during experimental procedure
  • 12V low voltage power input
  • Two sizes to meet your application volume demands
  • Convenient and easy-to-read digital display
  • Small footprint
  • Lightweight design
  • Single (S) model can hold a full block, 2 half blocks, 4 quarter blocks or 1 half and 2 quarter blocks
  • Double (D) model can hold 2 full blocks, 4 half blocks, 8 quarter blocks or a combination of full/half and quarter blocks.
Applications:
  • Sample preparation
  • Enzyme preservation
  • Enzyme-substrate reactions
  • DNA amplification
Required Accessory:
  • Unit must be ordered with block(s)
TRUSTED_SUSTAINABILITY
Certifications/Compliance CE, cTUVus, RoHS
Controller Type Digital
Plug Type CN, EU, UK
Temperature Range (Metric) Ambient +2° to 100°C
Type Dry Bath/Block
Heating Rate 20 min. average heating rate (30° to 100°C, ambient at 25°C, with one 0.2mL block)
Includes Dry bath, CN, EU,and UK style plugs, block lifter, plastic lid
Model Compact Dry Bath D
No. of Blocks 2 full blocks, 4 half blocks, 8 quarter blocks or a combination of full/half and quarter blocks
Temperature Accuracy ≤ ± 0.5°C
Temperature Uniformity ≤ ± 0.5°C
Warranty 2 Year, Replacement
Dimensions (L x W x H) 200 x 200 x 25 mm (7.9 x 7.9 x 1 in.)
Length (English) 7.9 in.
Length (Metric) 200 mm
Width (English) 7.9 in.
Width (Metric) 200 mm
Height (English) 1 in.
Height (Metric) 25 mm
Weight (English) 3.3 lb.
Weight (Metric) <1.5 kg
Electrical Requirements 12 V, DC, 7 A
Voltage 100/240 V
Show More Show Less
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit