Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HDS Reusable Comfort Face Masks
Mask constructed of antimicrobial fabric with lab results of 99.9% efficacy against bacteria hazardous to upper respiratory health. Speed Tech fabric has fluid repellancy and moisture wicking, allowing for an effective and comfortable mask.
Specifications
Product Type | General Purpose Face Mask |
---|---|
Fastening Type | Earloops |
Material | Polyester |
Size | Standard |
Sterility | Non-sterile |
Catalog Number | Color | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Color | Price | Quantity & Availability | ||||||
12-888-101
|
Black | Item Discontinued This item has been discontinued by Fisher Scientific. Please Sign In to view product availability in your area. | N/A | ||||||
12-888-102
|
White | Item Discontinued This item has been discontinued by Fisher Scientific. Please Sign In to view product availability in your area. | N/A | ||||||
Description
- Antimicrobial (Microban Z06)
- Zinc additive
- Machine washable, tumble dry
- Efficacy after 50 washes is 99% against 4352 KLEB and 6538 STAPH
- Water repellency
- Protects against coughs, sneezes, and other bodily fluids
- Moisture wicking
- Combats the condensation build up from breathing
- Non-stretch fabric
Specifications
General Purpose Face Mask | |
Polyester | |
Non-sterile | |
Yes |
Earloops | |
Standard | |
No |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title