Learn More
Greiner Bio-One ComboPlate™ CrystalBridge Protein Crystallization Plate
Ideal for protein crystallization. Greiner Bio-One ComboPlate™ CrystalBridge Protein Crystallization Plate is used for sitting drop applications.
Supplier: Greiner Bio-One 662145
Description
- Virgin, high-clarity polystyrene combined with distortion-free bottom provides optical properties for microscopic inspection of crystal growth
- Flattened raised ring around each well reduces risk of cross-contamination
- Raised protection lid protects cover slips and sealer during transportation and storage
- Conical well bottom
Specifications
Clear | |
Non-sterile | |
Well: 4.6 mm |
Polystyrene | |
250/Cs. |