Learn More
Metrohm USA COMBO GLASS ELECTRODE
Supplier:  Metrohm USA 60210100
Encompass
Combined pH glass electrode for sampler changers, SGJ, Metrohm plug-in head G. Diaphragm: Ceramic pin. Electrode plug-in head: Metrohm plug-in head G. Electrode slope: > 0.97. Electrode zero point (mV): 0 plus or minus 15 mV (pH = 7). Indicator electrode shape: Hemisphere. Indicator electrode type: Glass electrode. Internal reference electrode type: Ag wire + AgCl. Measuring unit: pH.
Catalog No. NC0442699