Learn More
Liofilchem™ ComASP Colistin / Piperacillin-tazobactam
The system is used to perform the broth microdilution (BMD) method for the antimicrobial susceptibility testing of colistin (CS) and piperacillin-tazobactam (TZP) as recommended by international standards (i.e. CLSI, EUCAST,ISO) but in a simpler and less time-consuming way.
Supplier: Liofilchem™ 75003
Alert:
Non-hazardous according to current legislation but fall within the specific field of application where a safety data sheet must be supplied because they can cause phenomena of sensitisation in sensitive subjects if they come into contact with the skin.
Description
- 4 Systems (panels) of ComASP Colistin/Pip-tazo (panels individually packed in foil with silica gel desiccant)
- 4 Vials of Mueller Hinton II Broth (7.2 mL)
Specifications
Colistin/Piperacillin-tazobactam | |
Store ComASP™ Colistin/Pip-tazo at 2°C to 8°C in the original packaging. Keep away from direct sunlight and direct heat. Do not use the panels beyond the expiry date indicated on the label. Eliminate without using if there are signs of deterioration | |
Non-sterile | |
4 Systems (panels) |
0.008 to 128 μg/mL | |
Panel | |
Media Testing |