Learn More
MilliporeSigma™ Supelco™ columnSaver Precolumn Filter
The new Supelco columnSaver offers all of the protection of conventional precolumn filters at much less the cost. With its convenient direct connect design, changeover time is measured in seconds, and requires no wrenches or tools to install.
$324.00 - $334.00
Specifications
Description 10/Pk.
View More Specs
Products 2
Catalog Number Mfr. No. Size (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Size (Metric) Price Quantity & Availability  
11-102-0792
View Documents
MilliporeSigma™ Supelco™
55214U
0.5 μm
Each for $324.00
Add to Cart
11-102-0791
View Documents
MilliporeSigma™ Supelco™
55215U
2.0 μm
Each for $334.00
Add to Cart
Description
Description
  • Economical protection for your guard or analytical HPLC column
  • No wrenches or tools required to install, fingertight to 5,000 psi
  • Lower dead volume than conventional filters with holders
  • Universal connection is compatible with all manufacturers fittings
The PEEK filter body contains a HiFlo filter element and is designed for maximum filtration of particulate matter with mimimal dead volume or backpressure. As soon as an increase in backpressure is observed, simply remove and dispose of the Supelco columnSaver and install a new one. The direct connect design is compatible with all 1/16 in.,10-32 internal fitting ports regardless of the manufacturer.
Specifications
Specifications
10/Pk.