Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Supelco™ Column Tags
For a permanent record of important chromatographic data
$51.75 - $144.00
Specifications
For Use With (Application) | Labeling/Marking Chromatography Columns |
---|---|
For Use With (Equipment) | Burgess Vibrograver, Scribing Tools |
Material | Aluminum |
Color | Silver |
Catalog Number | Mfr. No. | Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Type | Price | Quantity & Availability | |||||
11-102-0886
|
MilliporeSigma™ Supelco™
20396 |
Engraved |
Each for $144.00
|
|
|||||
11-102-0944
|
MilliporeSigma™ Supelco™
20401 |
Plain |
Each for $51.75
|
|
|||||
Specifications
Labeling/Marking Chromatography Columns | |
Burgess Vibrograver, Scribing Tools | |
Aluminum | |
Silver |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title