Learn More
BioVendor, LLC Empty EndoTrap Columns, Small (5 pack), for 0.2-2 ml resin
SDP
Supplier:  BioVendor, LLC LET0020
Encompass
Empty EndoTrap Columns, Small (5 pack), for 0.2-2 ml resin. Polypropylene. 0.3 inch diameter. 50 micron porous polyethylene discs. Accessory for use with EndoTrap red or EndoTrap HD Endotoxin Removal Kits by LIONEX. Affinity chromatography for highly efficient endotoxin removal. EndoTrap removes endotoxins (lipopolysaccharides/LPS) from aqueous solutions of a wide range of samples such as proteins, antibodies, vaccines, nucleic acids, buffers and cell extracts with excellent sample recovery rate. Helps avoid artefacts caused by endotoxin contamination in sensitive in-vitro and in-vivo assays. Fulfills requirements for industrial applications and can be easily integrated in early or late stage purification processes. Can be employed in both column or batch mode, by gravity flow or on fully automated liquid chromatography systems.
Catalog No. NC1802981