Learn More
Thermo Scientific Pre-Column Heater, 2μL ID, 0.13m
Supplier: Thermo Scientific 6722.0530
2 microliter internal diameter and 0.13 meter length pre-column heating device for liquid chromatography systems heats mobile phase before analytical column to control retention times, improve separation, reduce mobile phase viscosity, and decrease analyte interactions before separation.