Learn More
Epredia™ Colormark™ Slide Cartridges forSHUR/Mark™ Labeling Systems
Background color makes etched patient information more visible
$3.02 - $274.00
Catalog Number | Mfr. No. | Color | Coating | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Color | Coating | Price | Quantity & Availability | ||||
22-035815
|
Epredia™
CM5951W001 |
Blue | Non-adhesion |
Pack of 144 for $149.00
Case of 10 PK for $1,267.00
|
|
||||
22-038325
|
Epredia™
CM5951WPLUS001 |
Blue | Adhesion |
Pack of 144 for $274.00
Case of 10 PK for $2,312.00
|
|
||||
22-038328
|
Epredia™
CM9951WLPLUS001 |
Lilac/Black | Adhesion |
Pack of 144 for $266.00
Case of 10 PK for $2,232.00
|
|
||||
22-035817
|
Epredia™
CM7951W001 |
Pink | Non-adhesion |
Pack of 144
for $3.02
Save 98% Reg :
Case of 10 PK
for $30.18
Save 97% Reg : |
|
||||
22-038323
|
Epredia™
CM7951WPLUS001 |
Pink | Adhesion |
Pack of 144 for $273.00
Case of 10 PK for $2,312.00
|
|
||||
22-035814
|
Epredia™
CM4951W001 |
White | Non-adhesion |
Pack of 144 for $154.00
Case of 10 PK for $1,312.00
|
|
||||
22-038324
|
Epredia™
CM4951WPLUS001 |
White | Adhesion |
Pack of 144 for $274.00
Case of 10 PK for $2,312.00
|
|
||||
22-035818
|
Epredia™
CM8951W001 |
Yellow | Non-adhesion |
Pack of 144 for $154.00
Case of 10 PK for $1,312.00
|
|
||||
22-038327
|
Epredia™
CM8951WPLUS001 |
Yellow | Adhesion |
Pack of 144 for $273.00
Case of 10 PK for $2,312.00
|
|
||||
22-035816
|
Epredia™
CM6951W001 |
Green | Non-adhesion |
Item Discontinued
This item has been discontinued by the supplier. Please Sign In to view product availability in your area. View alternate product |
N/A | ||||
22-038326
|
Epredia™
CM6951WPLUS001 |
Green | Adhesion | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | ||||
Description
- Brightly coated with contrasting background color
- Self-contained packages ready for loading into your SHUR/Mark slide labeling system
- Simply remove the cover from the slide package and drop the sleeve into the loading bin of the SHUR/Mark labeling system
- Available with Plus formula to provide the utmost adhesion
Specifications
144 Pk., 10 Pk./Cs. |