Learn More
Color Standard, Pt-Co, 500 Units APHA, Stock Solution, Certified, LabChem™
$236.50 - $985.50
Specifications
Chemical Name or Material | Color Standard |
---|---|
Name Note | Pt-Co, 500 Units APHA, Stock Solution |
Packaging | Amber Glass |
CAS Max % | 95.43, 4.34, 0.13, 0.1 |
CAS | 16921-30-5, 7791-13-1, 7647-01-0, 7732-18-5 |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
LC133007
|
LabChem, Inc.
LC133007 |
125 mL | Amber Glass |
Each for $236.50
|
|
||||
LC133001
|
LabChem, Inc.
LC133001 |
500 mL | Amber Glass |
Each for $539.50
|
|
||||
LC133002
|
LabChem, Inc.
LC133002 |
1 L | Amber Glass |
Each for $985.50
|
|
||||
Specifications
Color Standard | |
Amber Glass | |
16921-30-5, 7791-13-1, 7647-01-0, 7732-18-5 | |
K2(PtCl6) | |
485.99 | |
Passes Test | |
Liquid |
Pt-Co, 500 Units APHA, Stock Solution | |
95.43, 4.34, 0.13, 0.1 | |
Orange | |
UN1789 | |
Certified | |
Soluble in water |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause cancer.
May damage fertility or the unborn child.
GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Do not breathe mist, vapors, spray.
Contaminated work clothing must not be allowed out of the workplace.
Wear protective gloves, eye protection, protective clothing, face protection.
Wear respiratory protection.
Wash exposed skin thoroughly after handling.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediatel
Danger
Recommended Storage : Room Temperature
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.