Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Remco™ Color-Coded 24 in. Hazmat Handle
Remco™ Color-Coded 24 in. Hazmat Handle allows you to clean a chemical spill economically and efficiently.
Supplier: Remco™ 6024
Description
- Key elements of a low-cost, total solution to chemical clean-up tasks
- Stiff but lightweight, they feature honeycomb construction with smooth, chemically resistant gel-coat surfaces
- Color-coded
Specifications
Yes | |
Non-sterile | |
24 in. | |
Handle |
White | |
1.3 in. | |
1 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title