Learn More
Aldon Corporation Coliform Test Powder Kit
Coliform test powder kit is an extremely easy way to test your water sample for coliforms.
$4.60 - $16.00
Catalog Number | Mfr. No. | Class Size | Includes | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Class Size | Includes | Price | Quantity & Availability | ||||
S06785
|
Aldon Corporation
IS5055 |
1 Lab Group | Instructions, 1 Coliform Powder Test Kit |
Each for $4.60
|
|
||||
S06786
|
Aldon Corporation
IS5056 |
5 Lab Groups | Instructions and 5 Coliform Powder Test Kits |
Each for $16.00
|
|
||||
Description
- Simply add a small amount of collected water to the sample tube and incubate for 24 to 48 hours.
- A distinct color change from purple to yellow indicates that coliforms are present.
Specifications
Coliform Powder Test Tube | |
9 to 12 |