Learn More
Sigma Aldrich Fine Chemicals Biosciences ColiComplete, 100 tests
Supplier: Sigma Aldrich Fine Chemicals Biosciences 10800100
ColiComplete is a patented substrate supporting disc method containing both XGAL and MUG. It is designed to simultaneously detect Total Coliforms and E. coli in food products.