Learn More
MilliporeSigma Coli-Count Test Kit, 25/pk
Supplier: MilliporeSigma MCSK10025
The Coli-Count Swab Test Kit enables a fast and easy Coliform monitoring in food and beverages, dental unit water, dialysate, pharmaceuticals and cosmetics. This kit finds its application in Clinical Lab, Environmental Monitoring, Food and Beverage and Microbial monitoring.