Learn More
Cole-Parmer Essentials Graduated Cylinder, PP 1000 mL; 3/PK
Supplier: Cole-Parmer 0613476
Bulk packed for greater savings, these polypropylene (PP) cylinders feature excellent transparency. Molded in one piece for enhanced durability, these cylinders have raised graduations for easy reading. Autoclavable.