Learn More
Cole-Parmer Cole-Parmer Burette and Burette Stand Kit; Burette, Clamp, and Stand
Supplier:  Cole-Parmer 2500055
Encompass_Preferred
Bundled for convenience to save you money, this Class B burette meets basic measuring and dispensing requirements. It is complete with a burette clamp and lab stand. This handy kit comes with a nickel-plated zinc and double burette clamp, along with a clamp holder and lab stand with aluminum rod and chemical-resistant porcelain base. The burette clamp can hold up to two burettes. The kit comes with a 50-mL burette.
Catalog No. NC1642715