Learn More
Yamato Scientific America, Inc. Yamato 4L Cold Trap, Max low temp of -85°C, 115V/220V.
Supplier: Yamato Scientific America, Inc. CA801
4L cold trap that efficiently traps water vapor and toxic substances discharged from vacuum oven and rotary evaporator to protect vacuum pump. Excellent choice to extract acid and organic solvents with the optional glass condenser. Efficiently reduces vapor inhalation amount to the vacuum pump. Can be used as a low temperature tank as well as pre-cooling tank. Utilizes R404A. Space saving and highly mobile on wheels. Max. low temp -85°C. Dehumidifying capacity max 1.0 kg.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.