Learn More
Andwin Scientific COLD PLATES 6/PKG
Supplier: Andwin Scientific 25608942
The Sakura Finetek Tissue-Tek Cold Plate is ideal for chilling and preparing paraffin blocks for microtome sectioning. This product can also be utilized as a cold tray for other lab procedures. It only takes 4 hours to be freezed and its surface will remain cold for up to 3 hours. This is sold in a pack of 6.