Learn More
Chemglass Life Sciences Condenser, Micro Cold Finger, 14/20 Inner Joint, 13mm Length Below Joint
Supplier: Chemglass Life Sciences CG1222A02
Condenser, Micro Cold Finger, 14/20 Inner Joint, 13mm Length Below Joint. Condenser for use in refluxing from 1 to 20mL of solvent in flasks having a capacity of 5 to 50mL. Minimal surface area provides for better yields while providing adequate cooling. The additional hose connection is provided to allow introduction of an inert gas such as nitrogen. Top hose connection is cooling inlet and middle hose connection is cooling outlet. Cold finger extends either 5mm or 13mm below the joint and is used on a flask with the listed joint size. Hose connection is 10mm at the largest serration. Design suggested by Dr. Kathleen Zandi, Pfizer Inc., Groton CT.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.