Learn More
Thermo Scientific™ Cold Filterable Tryptone Soya Broth in BPCs
Minimize prep time and maximize production for media fill trials with this ready-to-use soya broth in BioProcess Containers.
$802.00 - $4075.00
Specifications
Color | Clear, straw |
---|---|
Content And Storage | 2 to 25°C |
Description | Cold Filterable Tryptone Soya Broth in BPCs |
Format | Bioprocess Container |
Product Type | Microbiology Media |
Catalog Number | Mfr. No. | dimFormat | Quantity | Includes | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | dimFormat | Quantity | Includes | Price | Quantity & Availability | |||
BP1065E
|
Thermo Scientific™
BP1065E |
20 L | Includes the following connectors: SmartSite, needle-free valve additional sampling port, Quick connect, +ö body (MPC), 1+ö triclamp and ReadyMate aseptic connector |
Each for $4,075.00
|
|
||||
BP1065A
|
Thermo Scientific™
BP1065A |
1 L | Includes the following connectors: LUER lock female +ö and SmartSite, needle-free valve additional sampling port |
Each for $802.00
|
|
||||
BP1065C
|
Thermo Scientific™
BP1065C |
10 L | Includes the following connectors: SmartSite, needle-free valve additional sampling port, Quick connect, +ö body (MPC), 1+ö triclamp and ReadyMate aseptic connector |
Each for $3,027.00
|
|
||||
Description
Minimize prep time and maximize production for media fill trials with ready-to-use Thermo Scientific™ Cold Filterable Tryptone Soya Broth in BPCs. Simply connect the BioProcess Containers into your existing workflow to maximize process simulations and minimize secondary contamination.
Effective- Achieve effective process simulations, with minimized risk of secondary contamination
- Just plug and play
- Minimize preparation time
- Satisfy regulatory compliance
- Save time, reduce labour costs
- Innovative bag design and connector options for easy process integration
- Standard, ready-to-use, off-the-shelf product range
- No mixing, no sterilization, long shelf life at ambient storage
- No Mycoplasma contamination
- Qualified microbial recovery performance
- Comprehensive validation package for media, bag/components and filling process
- No filter blockages
- No mixing required
- No sterilization
- No WFI (water for injection) required
- No vessel cleaning
Specifications
Clear, straw | |
Cold Filterable Tryptone Soya Broth in BPCs | |
Microbiology Media |
2 to 25°C | |
Bioprocess Container |
Safety and Handling
ShelfLife : 2 Years
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.